Anti-CRISPR-Cas9 Mouse Monoclonal Antibody [clone: 7A9-3A3]

Supplier: Novus Biologicals
NBP2-36440
MSPP-NBP236440EA 564.92 USD
MSPP-NBP236440
Anti-CRISPR-Cas9 Mouse Monoclonal Antibody [clone: 7A9-3A3]
Antibodies
The CRISPR-Cas9 Antibody (7A9-3A3) - N-Terminus from Novus Biologicals is a mouse monoclonal antibody to CRISPR-Cas9. This antibody reacts with bacteria. The CRISPR-Cas9 Antibody (7A9-3A3) - N-Terminus has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount.

Type: Primary
Antigen: CRISPR-Cas9
Clonality: Monoclonal
Clone: 7A9-3A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR