Blunt Tip Needles, Braintree Scientific
Supplier: BRAINTREE SCIENTIFIC INC MS
NB-20GC
MSPP-NB20GCPK 224.62 USD
MSPP-NB20GC
Blunt Tip Needles, Braintree Scientific
Syringe Needles
Stainless steel, blunt tip, hypodermic needle with standard Luer hub.
For use as an interface to connect a hypodermic syringe directly to catheter port for intravascular substance administration, blood sampling and catheter flushing.