Mouse Tail Illuminator Restrainer, Braintree Scientific INC
Supplier: 0000018294
MTI-STD MTI-TUBE-3
MSPP-MTISTDEA 692.31 USD
MSPP-MTISTD MSPP-MTITUBE3
Mouse Tail Illuminator Restrainer, Braintree Scientific INC
Animal Supplies Animal Restraints
These mouse tail illuminator restrainers efficiently illuminates and warms a mouse tail for easy injection or bleeding.
  • Restraining tube slides off base for easy cleaning
  • Illuminated slot for easy tail vein injection
Features sliding doors to keep mouse gently restrained.
Ordering information: Additional tubes are available and should be ordered separately.
Delivery information: Each unit comes with a restraining tube and illumination source.