MonoSLEEVE™ Controller
Supplier: ANALYTICAL SALES & SERVICE MS
MONOSLEEVE
MSPP-MONOSLEEVEA 2376.92 USD
MSPP-MONOSLEEV
MonoSLEEVE™ Controller
Controllers Temperature Controllers
MonoSLEEVE is the next generation of the original Hot Sleeve controller. With its new advanced “Plug & Play” technology, MonoSleeve can accommodate single HotSleeve+* or AgileSleeve+®* column heaters of different sizes without having to recalibrate to each individual sleeve. This allows you to switch out different heaters on the fly.
Delivery information: Includes: MonoSLEEVE Controller, Power Supply, USB Cable, GUI, and Manual. Heater not included.
Caution: The temperature controller must be operated away from liquids so as not to accidentally spill solvents on the top cover. Do not immerse or operate any part of the Column Heater in liquids. In the event of solvent leakage, wipe sleeve clean before further use.