Mouse Dissection Surgical Kit, Braintree Scientific
Supplier: BRAINTREE SCIENTIFIC INC MS
MDK-M
MSPP-MDKMEA 238.46 USD
MSPP-MDKM
Mouse Dissection Surgical Kit, Braintree Scientific
Dissection Sets
Mouse dissection kit with storage case.
Delivery information: Includes: Dumont Forceps #5, Vannas Scissor, Dissecting Scissors, Wire Retractor, Needle Holder and Blunt Probe.