Anti-COL3a1 Mouse Monoclonal Antibody [clone: C24]

Supplier: CLOUD-CLONE CORP MS
MAD195RA21
MSPP-MD195RA21EA 400 USD
MSPP-MD195RA21
Anti-COL3a1 Mouse Monoclonal Antibody [clone: C24]
Antibodies
Monoclonal antibody to Collagen Type III Alpha 1 (COL3a1), derived from recombinant COL3a1(Arg1171~Arg1435), is reactive with Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Collagen alpha-1(III) chain is a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I collagen. Mutations in this gene are associated with Ehlers-Danlos syndrome type IV, and with aortic and arterial aneurysms. Although alternate transcripts have been detected for this gene, they are the result of mutations, these mutations alter splicing, often leading to the exclusion of multiple exons. By fluorescence in situ hybridization, Limongi et al. (1997) concluded that the COL3A1 gene is located in chromosome band 2q32.2 and that the nebulin gene is located in band 2q24.2, the FRA2G fragile site was found to lie between the 2 genes in the 2q31 band.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: COL3a1
Clonality: Monoclonal
Clone: C24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR