Anti-UCN3 Mouse Monoclonal Antibody [clone: C9]

Supplier: CLOUD-CLONE CORP MS
MAD140HU21
MSPP-MD140HU21EA 370 USD
MSPP-MD140HU21
Anti-UCN3 Mouse Monoclonal Antibody [clone: C9]
Antibodies
Monoclonal Antibody to Urocortin 3 (UCN3), derived from recombinant UCN3 (Leu22~Lys161), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Urocortin-3 is a member of the sauvagine/corticotropin-releasing factor/urotensin I family. It is structurally related to the corticotropin-releasing factor (CRF) gene and the encoded product is an endogenous ligand for CRF type 2 receptors. In the brain it may be responsible for the effects of stress on appetite. In spite of the gene family name similarity, the product of this gene has no sequence similarity to urotensin II. The mature human SCP peptide sequence shares 72% identity with that of Takifugu rubripes and 57% identity with that of Tetraodon nigroviridis, a freshwater pufferfish. In human and fish, a stretch of 30 residues at the respective carboxy termini has an extended alpha-helical structure shared by all CRH family peptides.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: UCN3
Clonality: Monoclonal
Clone: C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR