Anti-COL4a5 Mouse Monoclonal Antibody [clone: 7#]

Supplier: CLOUD-CLONE CORP MS
MAC141HU21
MSPP-MC141HU21EA 352 USD
MSPP-MC141HU21
Anti-COL4a5 Mouse Monoclonal Antibody [clone: 7#]
Antibodies
Monoclonal antibody to Collagen Type IV Alpha 5 (COL4a5), derived from recombinant COL4a5(Gly1461~Thr1685), is reactive with Human/Mouse/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

COL4a5 is one of the six subunits of type IV collagen, the major structural component of basement membranes. Mutations in this gene are associated with X-linked Alport syndrome, also known as hereditary nephritis. Like the other members of the type IV collagen gene family, this gene is organized in a head-to-head conformation with another type IV collagen gene so that each gene pair shares a common promoter. Three transcript variants have been identified for this gene. Type IV collagen is the major structural component of glomerular basement membranes (GBM), forming a 'chicken-wire' meshwork together with laminins, proteoglycans and entactin/nidogen.There are six type IV collagen isoforms, alpha 1(IV)-alpha 6(IV), each of which can form a triple helix structure with 2 other chains to generate type IV collagen network.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: COL4a5
Clonality: Monoclonal
Clone: 7#
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR