Anti-GDF11 Mouse Monoclonal Antibody [clone: C4]

Supplier: CLOUD-CLONE CORP MS
MAC113HU21
MSPP-MC113HU21EA 370 USD
MSPP-MC113HU21
Anti-GDF11 Mouse Monoclonal Antibody [clone: C4]
Antibodies
Monoclonal Antibody to Growth Differentiation Factor 11 (GDF11), derived from recombinant GDF11 (Thr138~Asn386), is reactive with Human/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

GDF11 is a protein belonging to the transforming growth factor beta superfamily that controls anterior-posterior patterning by regulating the expression of Hox genes.It determines Hox gene expression domains and rostrocaudal identity in the caudal spinal cord. During mouse development, GDF11 expression begins in the tail bud and caudal neural plate region. This cytokine also inhibits the proliferation of olfactory receptor neuron progenitors to regulate the number of olfactory receptor neurons occurring in the olfactory epithelium, and controls the competence of progenitor cells to regulate numbers of retinal ganglionic cells developing in the retina. GDF11 can bind type I TGF-beta superfamily receptors ALK4, ALK5 and ALK7), but predominantly uses ALK4 and ALK5 for signal transduction.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: GDF11
Clonality: Monoclonal
Clone: C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR