Anti-APOA4 Mouse Monoclonal Antibody [clone: A8]

Supplier: CLOUD-CLONE CORP MS
MAB967PO21
MSPP-MB967PO21EA 420 USD
MSPP-MB967PO21
Anti-APOA4 Mouse Monoclonal Antibody [clone: A8]
Antibodies
Monoclonal Antibody to Apolipoprotein A4 (APOA4), derived from recombinant APOA4 (Glu21~Asn235), is reactive with Human/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

APOA4 resides on chromosome 11 in close linkage to APOA1 and APOC3. APOA4 contains 3 exons separated by two introns, and is polymorphic, although most of the reported sequence polymorphisms occur in exon 3. The best validated and studied non-synonymous SNPs are a glutamine histidine substitution at codon 380 and a threonine serine substitution at codon 367; a sequence polymorphism has also been identified in the 3'UTR of the third exon. Intra-species comparative gene sequence analysis suggests that the APOA4 gene arose from APOA1 by gene duplication approximately 270 MYA.The primary translation product of the APOA4 gene is a 396-residue preprotein, which undergoes proteolytic processing to yield apo A-IV, a 376-reside mature O-linked glycoprotein.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: APOA4
Clonality: Monoclonal
Clone: A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR