Anti-LPS Mouse Monoclonal Antibody [clone: C6]

Supplier: CLOUD-CLONE CORP MS
MAB526GE21
MSPP-MB526GE21EA 398 USD
MSPP-MB526GE21
Anti-LPS Mouse Monoclonal Antibody [clone: C6]
Antibodies
Monoclonal Antibody to Lipopolysaccharide (LPS), derived from OVA conjugated LPS, is reactive with General species.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

LPS is a major component of the outer membrane of Gram-negative bacteria, contributing greatly to the structural integrity of the bacteria, and protecting the membrane from certain kinds of chemical attack. LPS also increases the negative charge of the cell membrane and helps stabilize the overall membrane structure. It is of crucial importance to gram negative bacteria, whose death results if it is mutated or removed., LPS is an endotoxin, and induces a strong response from normal animal immune systems.LPS acts as the prototypical endotoxin because it binds the CD14/TLR4/MD2 receptor complex, which promotes the secretion of pro-inflammatory cytokines in many cell types, but especially in macrophages. In Immunology, the term 'LPS challenge' refers to the process of exposing a subject to an LPS which may act as a toxin.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: LPS
Clonality: Monoclonal
Clone: C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR