Anti-5-LO Mouse Monoclonal Antibody [clone: D10]

Supplier: CLOUD-CLONE CORP MS
MAB355HU21
MSPP-MB355HU21EA 352 USD
MSPP-MB355HU21
Anti-5-LO Mouse Monoclonal Antibody [clone: D10]
Antibodies
Monoclonal Antibody to 5-Lipoxygenase (5-LO), derived from recombinant 5-LO(Glu461~Ser671), is reactive with Human/Mouse/Rat/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Arachidonate 5-lipoxygenase is a member of the lipoxygenase family. It transforms EFAs into leukotrienes and is a current target for pharmaceutical intervention in a number of diseases.5-ALOX catalyzes oxidation of AA at the 5-position to yield 5-HPETE. 5-LO then converts 5-HPETE to leukotriene A4.5-ALOX is a target for pharmaceutical intervention in CAD.,Some people with variant alleles for 5-LO are at elevated risk for CAD. 5-ALOX is expressed in brain cells and may participate in neuropathologic processes.Mutations in the promoter region of this gene lead to a diminished response to antileukotriene drugs used in the treatment of asthma and may also be associated with atherosclerosis and several cancers. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: 5-LO
Clonality: Monoclonal
Clone: D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human;Mouse;Rat;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR