Anti-ACTa2 Mouse Monoclonal Antibody [clone: F1]

Supplier: CLOUD-CLONE CORP MS
MAB342MI21
MSPP-MB342MI21EA 386 USD
MSPP-MB342MI21
Anti-ACTa2 Mouse Monoclonal Antibody [clone: F1]
Antibodies
Monoclonal Antibody to Actin Alpha 2, Smooth Muscle (ACTa2), derived from recombinant ACTa2 (Ile167~Phe377), is reactive with Human/Mouse/Rat/Dog/Pig/Bovine.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

ACTA2 encodes the protein actin, alpha 2, which is a major component of the contractile apparatus in smooth muscle cells. This protein is crucial for muscle contraction and is involved in various cellular processes, including maintaining cell shape, motility, and division. ACTA2 is highly expressed in vascular smooth muscle cells and is essential for the contractile function of blood vessels. Mutations in the ACTA2 gene are linked to several vascular disorders, including thoracic aortic aneurysms and dissections. It is also a marker for smooth muscle differentiation and is used in research to study smooth muscle cell function and related diseases.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: ACTa2
Clonality: Monoclonal
Clone: F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse;Rat;Dog;Pig;Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR