Thermoshaker for Microplates
Supplier: BIOTANG INC. MS
MB100-4A
MSPP-MB1004AEA 3615.38 USD
MSPP-MB1004A
Thermoshaker for Microplates
Shakers and Mixers Shaking Incubators
The MB100-4A is designed for shaking 1 to 4 standard 96-well plates in the thermostating mode.
  • Microprocessor controlled time, RPM, and temperature
  • Heating plates above and below the microplates
  • Incubation up to 70 °C in 0.1 °C steps
  • Shakes and incubates four microplates simultaneously
  • Beep-signal and stop of shaking motion after program completion
  • Excellent temperature accuracy and uniformity across the plate
It is designed using the multi-principle which allows using it as two independent devices:
1. Incubator for lasting incubation without shaking of micro-quantities (insect, plant cell cultures, etc.) in immunoplates.
2. Microplate thermo-shaker for immunochemistry and molecular diagnostics, where the requirements to the result reproducibility and thus to the precise method regulation are particularly high.
A distinctive feature for the MB100-4A is two-side heating of microtest plates that allows achieving exact correspondence of the set and actual temperature in wells of the microtest plates.
This device can be used in
1. Cytochemistry - for in situ reactions.
2. Immunochemistry - for immunofermentative reactions.
3. Biochemistry - for enzyme and protein analysis.
4. Molecular chemistry - for matrix analysis.