Anti-Ang1-7 Mouse Monoclonal Antibody [clone: A4]

Supplier: CLOUD-CLONE CORP MS
MAS085MI21
MSPP-MAS85MI21EA 438 USD
MSPP-MAS85MI21
Anti-Ang1-7 Mouse Monoclonal Antibody [clone: A4]
Antibodies
Monoclonal Antibody to Angiotensin 1-7 (Ang1-7), derived from recombinant Ang1-7(Asp34~Pro40), is reactive with Human/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Angiotensin(1-7) (Ang1-7) acting at the level of the rostral ventrolateral medulla (RVLM) affects arterial pressure. Astroglia are a plausible cellular target of Ang1-7 in RVLM. Astrocytic responsiveness to Ang1-7 is reduced in SHR. Ang1-7 modulates astrocytic signalling which in vivo may affect local metabolism and microcirculation, resulting in changes in activity of RVLM pre-sympathetic neurones and hence blood pressure.,Ang1-7 is an important component of the central renin-angiotensin system and affects haemodynamics when injected into the RVLM. Effect of Ang1-7 could be blocked by co-application of its antagonist, A779 (200 nM) but not losartan (1 µM). non-CA neurones and glia in RVLM are the primary targets for Ang1-7 and its effect on [Ca2+]i in non-CA neurones is enhanced in SHR.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: Ang1-7
Clonality: Monoclonal
Clone: A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Lambda
Reactivity: Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR