Anti-OSCAR Mouse Monoclonal Antibody [clone: C2]

Supplier: CLOUD-CLONE CORP MS
MAC695HU22
MSPP-MAC695HU2EA 370 USD
MSPP-MAC695HU2
Anti-OSCAR Mouse Monoclonal Antibody [clone: C2]
Antibodies
Monoclonal Antibody to Osteoclast Associated Receptor (OSCAR), derived from recombinant OSCAR (Pyr~Gly95~Pro109), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Osteoclasts are multinucleated cells that resorb bone and are essential for bone homeostasis. This gene encodes an osteoclast-associated receptor (OSCAR), which is a member of the leukocyte receptor complex (LRC) protein family that plays critical roles in the regulation of both innate and adaptive immune responses. Different from the other LRC members, OSCAR expression is detected specifically in preosteoclasts or mature osteoclasts., Osteoclast-associated immunoglobulin-like receptor may be an important bone-specific regulator of osteoclast differentiation. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: OSCAR
Clonality: Monoclonal
Clone: C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR