Anti-APOB Mouse Monoclonal Antibody [clone: C3]

Supplier: CLOUD-CLONE CORP MS
MAC003RA21
MSPP-MAC03RA21EA 400 USD
MSPP-MAC03RA21
Anti-APOB Mouse Monoclonal Antibody [clone: C3]
Antibodies
Monoclonal Antibody to Apolipoprotein B (APOB), derived from recombinant APOB (H+Phe2747~Leu2913), is reactive with Rat/Human/Mouse/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Apolipoprotein B (APOB) is the primary apolipoprotein of low-density lipoproteins (LDL or 'bad cholesterol'), which is responsible for carrying cholesterol to tissues. While it is unclear exactly what functional role APOB plays in LDL, it is the primary apolipoprotein component and is absolutely required for its formation. What is clear is that the APOB on the LDL particle acts as a ligand for LDL receptors in various cells throughout the body., Through a mechanism that is not fully understood, high levels of APOB can lead to plaques that cause vascular disease (atherosclerosis), leading to heart disease. There is considerable evidence that levels of APOB are a better indicator of heart disease risk than total cholesterol or LDL.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: APOB
Clonality: Monoclonal
Clone: C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Rat;Human;Mouse;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR