Anti-ALB Mouse Monoclonal Antibody [clone: C1]

Supplier: CLOUD-CLONE CORP MS
MAB028HU22
MSPP-MAB028HU2EA 290 USD
MSPP-MAB028HU2
Anti-ALB Mouse Monoclonal Antibody [clone: C1]
Antibodies
Monoclonal Antibody to Albumin (ALB), derived from native ALB, is reactive with Human/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Albumin is a soluble, monomeric protein which comprises about one-half of the blood serum protein. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Mutations in the ALB gene on chromosome 4 result in various anomalous proteins. Albumin is a globular unglycosylated serum protein of molecular weight 65000. The albumin variant first described by Fraser et al. in a Welsh family was characterized as a dimer by Jamieson and Ganguly. The amino acid sequence has been determined in fragments of serum albumin of man. By 1980, at least 2 dozen electrophoretic variants of serum albumin had been reported but only 2 of them had been characterized with respect to their primary structure: albumin A and albumin B.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: ALB
Clonality: Monoclonal
Clone: C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR