Anti-MIP3a Mouse Monoclonal Antibody [clone: A4]

Supplier: CLOUD-CLONE CORP MS
MAA095HU21
MSPP-MAA95HU21EA 290 USD
MSPP-MAA95HU21
Anti-MIP3a Mouse Monoclonal Antibody [clone: A4]
Antibodies
Monoclonal antibody to Macrophage Inflammatory Protein 3 Alpha (MIP3a), derived from recombinant MIP3a(Ala27~Met96), is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Chemokine (C-C motif) ligand 20 is a small cytokine belonging to the CC chemokine family. It is strongly chemotactic for lymphocytes and weakly attracts neutrophils.CCL20 is implicated in the formation and function of mucosal lymphoid tissues via chemoattraction of lymphocytes and dendritic cells towards the epithelial cells surrounding these tissues. CCL20 elicits its effects on its target cells by binding and activating the chemokine receptor CCR6.CCL20 can be induced by microbial factors such as lipopolysaccharide, and inflammatory cytokines such as tumor necrosis factor and interferon-γ, and down-regulated by IL-10. CCL20 is expressed in several tissues with highest expression observed in peripheral blood lymphocytes, lymph nodes, liver, appendix, and fetal lung and lower levels in thymus, testis, prostate and gut.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: MIP3a
Clonality: Monoclonal
Clone: A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR