Anti-PINP Mouse Monoclonal Antibody [clone: H20]

Supplier: CLOUD-CLONE CORP MS
MAA957HU22
MSPP-MAA957HU2EA 550 USD
MSPP-MAA957HU2
Anti-PINP Mouse Monoclonal Antibody [clone: H20]
Antibodies
Monoclonal Antibody to Procollagen I N-Terminal Propeptide (PINP), derived from recombinant PINP (Gln23~Pro161), is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The PINP molecule is similar to PIIINP consisting of three distinct structural domains: Col 1 is on the aminoterminal side of the molecule, while Col 2 and Col 3 are situated on the middle of the helically structured molecule (Kühn et al., 1982). The PINP molecule has a molecular mass of 35000 and is cleared by scavenger receptors in liver endothelial cells (Melkko et al., 1994). PINP often occurs in circulation in two forms of different molecular sizes. One is identical to the trimeric authentic antigen (intact PINP) whereas the other consists of smaller forms of PINP, resembling a single domain of the proα1(I) chain of PINP and is probably a degradation product of type I procollagen or I pN-collagen. Thus, an assay of intact PINP rather than total PINP appears to be more sensitive in detecting changes in the rate of type I collagen synthesis (Melkko et al., 1996, Risteli and Risteli 1999).

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PINP
Clonality: Monoclonal
Clone: H20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR