Anti-MPO Mouse Monoclonal Antibody [clone: C3]

Supplier: CLOUD-CLONE CORP MS
MAA601HU22
MSPP-MAA601HU2EA 290 USD
MSPP-MAA601HU2
Anti-MPO Mouse Monoclonal Antibody [clone: C3]
Antibodies
Monoclonal Antibody to Myeloperoxidase (MPO), derived from recombinant MPO(Val165~Leu258), is reactive with Human/Mouse/Rat/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Myeloperoxidase (MPO) is a hemoprotein that is abundantly expressed in polymorphonuclear leukocytes (neutrophils) and secreted during their activation. The presence of a peroxidase in the cytoplasmic granules of leukocytes was suggested at the beginning of 20th century but it was the early 1940s that it was purified for the first time.,Native MPO is a covalently bound tetrameric complex of two glycosylated alpha chains (MW 59 to 64 kDa) and two unglycosylated beta chains (MW 14 kDa) with total MW about 150 kDa and theoretical pI 9.2. MPO plays an important role in neutrophil microbicidal action through catalyzing chloride ion oxidation to hypochlorous acid, which is a potent antimicrobial agent.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: MPO
Clonality: Monoclonal
Clone: C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human;Mouse;Rat;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR