Anti-TM Mouse Monoclonal Antibody [clone: C10]

Supplier: CLOUD-CLONE CORP MS
MAA529HU22
MSPP-MAA529HU2EA 336 USD
MSPP-MAA529HU2
Anti-TM Mouse Monoclonal Antibody [clone: C10]
Antibodies
Monoclonal Antibody to Thrombomodulin (TM), derived from recombinant TM (Ala19~Ser515), is reactive with Human/Pig.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Thrombomodulin (TM) is the cell surface receptor for thrombin. When occupied, thrombomodulin converts thrombin from a procoagulant protein into the activator of Protein C. Once activated Protein C (APC) has been generated, thrombomodulin acts as a major anticoagulant through its ability to inactivate various blood factors (Va, VΙΙΙa, Xa and XΙΙΙa). In competing for thrombin binding, thrombomodulin inhibits the proteolytic effect of thrombin in its clotting of fibrinogen, the inactivation of Protein S and the induction of platelet aggregation. TM is an integral membrane glycoprotein resembling in structure the low-density lipoprotein (LDL) receptor. TM possesses several EGF repeats, of which numbers five and six are responsible for the high affinity binding of thrombin (Kd = 0.5 nM).

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: TM
Clonality: Monoclonal
Clone: C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR