Anti-GZMM Mouse Monoclonal Antibody [clone: D1]

Supplier: CLOUD-CLONE CORP MS
MAA431HU22
MSPP-MAA431HU2EA 346 USD
MSPP-MAA431HU2
Anti-GZMM Mouse Monoclonal Antibody [clone: D1]
Antibodies
Monoclonal antibody to Granzyme M (GZMM), derived from recombinant GZMM(Ile26~Ala257), is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Human natural killer (NK) cells and activated lymphocytes express and store a distinct subset of neutral serine proteases together with proteoglycans and other immune effector molecules in large cytoplasmic granules. These serine proteases are collectively termed granzymes and include 4 distinct gene products: granzyme A, granzyme B, granzyme H, and Met-ase, also known as granzyme M. By fluorescence in situ hybridization, Pilat et al. (1994) mapped the GZMM gene to chromosome 19p13.3. Interphase studies with 2 differentially labeled probes for GZMM and the azurocidin (AZU1), proteinase-3 (PRTN3), and elastase-2 (ELA2) gene cluster indicated that the GZMM gene is 200 to 500 kb from that gene cluster.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: GZMM
Clonality: Monoclonal
Clone: D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR