Anti-SDF1 Mouse Monoclonal Antibody [clone: C3]

Supplier: CLOUD-CLONE CORP MS
MAA122HU22
MSPP-MAA122HU2EA 290 USD
MSPP-MAA122HU2
Anti-SDF1 Mouse Monoclonal Antibody [clone: C3]
Antibodies
Monoclonal Antibody to Stromal Cell Derived Factor 1 (SDF1), derived from recombinant SDF1 (Pro23~Lys89), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

SDF-1 is small cytokine belonging to the chemokine family that is officially designated Chemokine (C-X-C motif) ligand 12 (CXCL12).SDF-1 is produced in two forms, SDF-1α/CXCL12a and SDF-1β/CXCL12b, by alternate splicing of the same gene. SDF-1β is synthesized as a 93 amino acid (aa) precursor that contains a 21 aa signal sequence and a 72 aa mature region. The mature molecule exhibits a typical three antiparallel β-strand chemokine-like fold. There are no potential N-linked glycosylation sites. N-terminal aa ’s 1-8 form a receptor binding site, while aa’s 1 and 2 (Lys-Pro) are involved in receptor activation. The C-terminus is likely associated with heparin binding. SDF-1β circulates and undergoes proteolytic processing. SDF-1β are reported to be a monomers at neutral pH and physiologic ionic strength.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: SDF1
Clonality: Monoclonal
Clone: C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR