Anti-IL4 Mouse Monoclonal Antibody [clone: C5]

Supplier: CLOUD-CLONE CORP MS
MAA077HU22
MSPP-MAA077HU2EA 290 USD
MSPP-MAA077HU2
Anti-IL4 Mouse Monoclonal Antibody [clone: C5]
Antibodies
Monoclonal Antibody to Interleukin 4 (IL4), derived from recombinant IL4(His25~Ser153), is reactive with Human/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Interleukin 4 is a pleiotropic cytokine produced primarily by activated T lymphocytes,mast cells and basophils . IL-4 has multiple immune response-modulating functions on a variety of cell types.,It is an important modulator of the differentiation of precursor T helper cells to the Th2 subset that mediates humoral immunity and modulates antibody production. In addition, IL-4 has also been shown to have anti-tumor activity both in vivoand in vitro.The biological effects of IL-4 are mediated by specific cell surface receptor complexes. One type of functional IL-4 receptor complex consists of the IL-4-binding subunit and a second chain, designated the common c chain because it has also been identified as a component of the receptor complexes for IL-4, IL-7, IL-9 and IL-15.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR