Anti-IL18 Mouse Monoclonal Antibody [clone: C10]

Supplier: CLOUD-CLONE CORP MS
MAA064HU22
MSPP-MAA064HU2EA 305 USD
MSPP-MAA064HU2
Anti-IL18 Mouse Monoclonal Antibody [clone: C10]
Antibodies
Monoclonal Antibody to Interleukin 18 (IL18), derived from recombinant IL18 (Arg94~Asp193), is reactive with Human/Rat/Pig.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

IL-18 is a 18 kDa novel cytokine which is identified as a costimulatory factor for production of interferon-γ (IFN- γ ) in response to toxic shock. It shares functional similarities with IL-12. IL-18 is synthesized as a precursor 24 kDa molecule without a signal peptide and must be cleaved to produce an active molecule. IL-1 ß converting enzyme (ICE, Caspase-1) cleaves pro-IL-18 at aspartic acid in the P1 position, producing the mature, bioactive peptide that is readily released from the cells. It has been reported that IL-18 is produced from Kupffer cells, activated macrophages, keratinocytes, intestinal epithelial cells, osteoblasts, adrenal cortex cells and murine diencephalon. Apart from its physiological role, IL-18 is also able to induce severe inflammatory reactions, which suggests its role in certain inflammatory disorders.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: IL18
Clonality: Monoclonal
Clone: C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR