Anti-CYCS Mouse Monoclonal Antibody [clone: C2]

Supplier: CLOUD-CLONE CORP MS
MAA594HU21
MSPP-MA594HU21EA 352 USD
MSPP-MA594HU21
Anti-CYCS Mouse Monoclonal Antibody [clone: C2]
Antibodies
Monoclonal Antibody to Cytochrome C (CYCS), derived from recombinant CYCS (Glu5~Lys100), is reactive with Human/Mouse/Rat/Guinea pig/Rabbit/Dog/Pig/Goat/Chicken.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Cytochrome c, or cyt c (horse heart: PDB 1HRC) is a small heme protein found loosely associated with the inner membrane of the mitochondrion. It is a soluble protein, unlike other cytochromes, and is an essential component of the electron transfer chain. It is capable of undergoing oxidation and reduction, but does not bind oxygen. It transfers electrons between Complexes III and IV. Cytochrome c is a highly conserved protein across the spectrum of species, found in plants, animals, and many unicellular organisms. This, along with its small size (molecular weight about 12000 daltons), makes it useful in studies of evolutionary divergence. Its primary structure consists of a chain of 100 amino acids.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: CYCS
Clonality: Monoclonal
Clone: C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human;Mouse;Rat;Guinea pig;Rabbit;Dog;Pig;Goat;Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR