Anti-IL1b Mouse Monoclonal Antibody [clone: C8]

Supplier: CLOUD-CLONE CORP MS
MAA563RB21
MSPP-MA563RB21EA 340 USD
MSPP-MA563RB21
Anti-IL1b Mouse Monoclonal Antibody [clone: C8]
Antibodies
Monoclonal Antibody to Interleukin 1 Beta (IL1b), derived from recombinant IL1b (Val118~Ser268), is reactive with Rabbit.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Interleukin-1 (IL-1) is one of the first cytokines ever described. Its initial discovery was as a factor that could induce fever, control lymphocytes, increase the number of bone marrow cells and cause degeneration of bone joints. At this time, IL-1 was known under several other names including endogenous pyrogen, lymphocyte activating factor, haemopoetin-1 and mononuclear cell factor, amongst others., It was around 1984-1985 when scientists confirmed that IL-1 was actually composed of two distinct proteins, now called IL-1α and IL-1β.These belong to a family of cytokines known as the interleukin-1 superfamily.Both IL-1α and IL-1β are produced by macrophages, monocytes and dendritic cells.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: IL1b
Clonality: Monoclonal
Clone: C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR