Anti-NSE Mouse Monoclonal Antibody [clone: C12]

Supplier: CLOUD-CLONE CORP MS
MAA537HU21
MSPP-MA537HU21EA 336 USD
MSPP-MA537HU21
Anti-NSE Mouse Monoclonal Antibody [clone: C12]
Antibodies
Monoclonal Antibody to Enolase, Neuron Specific (NSE), derived from recombinant NSE (Met1~Leu434), is reactive with Human/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The glycolytic enzyme enolase (2-phosph-D-glycerate hydrolyase) exists as several dimeric isoenzymes (aa, ab, ag and gg) composed of three distinct subunits, a, b, and g. Three isoenzymes are found in rat brain: aa, ag and gg. The ag and gg-enolase isoenzymes are also known as neuron­-specific enolase (NSE) as these isoenzymes initially were detected in neurons and neuronendocrine cells. The NSE levels are low in health and benign subjects. Elevated levels are commonly found in patients with malignant tumors with neuronendocrine differentiation, especially small cell lung cancer and neuroblasloma. NSE are reported to be useful diagnostic marker for lung cancer, neuroblastoma, melanoma, seminoma and in injury of central nervous system.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: NSE
Clonality: Monoclonal
Clone: C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR