Anti-NT-ProBNP Mouse Monoclonal Antibody [clone: F8]

Supplier: CLOUD-CLONE CORP MS
MAA485HU21
MSPP-MA485HU21EA 360 USD
MSPP-MA485HU21
Anti-NT-ProBNP Mouse Monoclonal Antibody [clone: F8]
Antibodies
Monoclonal Antibody to N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP), derived from recombinant NT-ProBNP (His27~Arg102), is reactive with Human/Pig.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Brain-type Natriuretic Peptide (BNP) is a nonglycosylated peptide that is produced predominantly by ventricular myocytes and belongs to the natriuretic peptide family. Plasma NT-proBNP is a marker for congestive heart failure, while mature BNP (aa 103 to 134) promotes vasodilation and fluid and sodium excretion. BNP is co-secreted along with a 76 amino acid N-terminal fragment (NT-proBNP) which is biologically inactive. BNP binds to and activates the atrial natriuretic factor receptors NPRA, and to a lesser extent NPRB, in a fashion similar to atrial natriuretic peptide (ANP) but with 10-fold lower affinity. The biological half-life of BNP, however, is twice as long as that of ANP, and that of NT-proBNP is even longer, making these peptides better targets than ANP for diagnostic blood testing.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: NT-ProBNP
Clonality: Monoclonal
Clone: F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR