Anti-Cor Mouse Monoclonal Antibody [clone: C2]

Supplier: CLOUD-CLONE CORP MS
MAA462GE21
MSPP-MA462GE21EA 400 USD
MSPP-MA462GE21
Anti-Cor Mouse Monoclonal Antibody [clone: C2]
Antibodies
Monoclonal Antibody to Cortisol (Cor), derived from OVA conjugated Cor, is reactive with General species.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Cortisol is the major glucocorticoid produced and secreted by the adrenal cortex. It affects the metabolism of protein, fat and carbohydrates; the maintenance of muscle and myocardial integrity; and the suppression of inflammatory and allergic activities. Production of cortisol from the adrenal cortex is dependent upon corticotrophin (ACTH), which is secreted by the anterior pituitary. The corticotrophin releasing factor (CRF) which regulates ACTH is secreted by the hypothalamus and is responsive to cortisol levels. Physical, psychological and surgical stress and diurnal variation will affect the rate of cortisol production. Corticosteroid-binding globulin and albumin bind approximately 90% of the cortisol secreted by the adrenal cortex. Bound cortisol circulates in an available but temporarily inactive state. The physiological activity of cortisol depends upon levels of the small fraction of circulating unbound cortisol.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: Cor
Clonality: Monoclonal
Clone: C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Lambda
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR