Anti-E2 Mouse Monoclonal Antibody [clone: C1]

Supplier: CLOUD-CLONE CORP MS
MAA461GE21
MSPP-MA461GE21EA 400 USD
MSPP-MA461GE21
Anti-E2 Mouse Monoclonal Antibody [clone: C1]
Antibodies
Monoclonal Antibody to Estradiol (E2), derived from OVA conjugated E2, is reactive with General species.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Estradiol is an estrogenic hormone, C₁₈H₂₄O₂, produced by the ovaries and used in treating estrogen deficiency. Estradiol is essential for maintaining normal female functions and is normally produced by the ovaries. After menopause, the ovaries decrease their production of this hormone., Estradiol is a sex hormone. Estradiol is the predominant sex hormone present in females; however, it is present in males, albeit at lower levels, as well. It represents the major estrogen in humans. Estradiol has not only a critical impact on reproductive and sexual functioning, but also affects other organs including the bones.Estradiol is produced not only in the gonads: In both sexes, precursor hormones, to be specific testosterone, are converted by aromatization to estradiol. In particular, fat cells are active to convert precursors to estradiol, and will continue to do so even after menopause.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: E2
Clonality: Monoclonal
Clone: C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR