Anti-CPP Mouse Monoclonal Antibody [clone: C1]

Supplier: CLOUD-CLONE CORP MS
MAA365HU28
MSPP-MA365HU28EA 360 USD
MSPP-MA365HU28
Anti-CPP Mouse Monoclonal Antibody [clone: C1]
Antibodies
Monoclonal Antibody to Copeptin (CPP), derived from recombinant CPP (Asp128~Leu144), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Copeptin, a stable peptide of the arginine vasopressin precursor, is elevated in hemorrhagic and septic shock. Arginine vasopressin (AVP) levels are increased in hemorrhagic and septic shock. Measurement of AVP levels has limitations due to its short half-life and cumbersome detection method. Copeptin is a more stable peptide derived from the same precursor molecule. C-Terminal Provasopressin (Copeptin) as a Novel and Prognostic Marker in Acute Myocardial Infarction. Arginine vasopressin-neurophysin II is a precursor protein consisting of arginine vasopressin and two associated proteins, neurophysin II and a glycopeptide, copeptin. Arginine vasopressin is a posterior pituitary hormone which is synthesized in the supraoptic nucleus and paraventricular nucleus of the hypothalamus.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: CPP
Clonality: Monoclonal
Clone: C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR