Anti-E1S Mouse Monoclonal Antibody [clone: C3]

Supplier: CLOUD-CLONE CORP MS
MAA191GE21
MSPP-MA191GE21EA 498 USD
MSPP-MA191GE21
Anti-E1S Mouse Monoclonal Antibody [clone: C3]
Antibodies
Monoclonal Antibody to Estrone Sulfate (E1S), derived from OVA conjugated E1S, is reactive with General species.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Estrone sulfate(E1S) is a sulfated estrone. It has been described as a prognosis indicator for prostate cancer. It is a component of Premarin. Estrone sulfate is present at high concentration in plasma. It is a major circulating plasma estrogen and has a long plasma half-life; the large contributions of estrone and estradiol to plasma E1S are more than sufficient to account for all the circulating plasma E1S. It has been described as a prognosis indicator for prostate cancer. It is a component of Premarin. E1S is present at high concentration in plasma. Using average production rates, all plasma E1S could be shown to be derived from plasma estrone and estradiol.E1S is a major circulating plasma estrogen and has a long plasma half-life; (b) the large contributions of estrone and estradiol to plasma E1S are more than sufficient to account for all the circulating plasma E1S.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: E1S
Clonality: Monoclonal
Clone: C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR