Anti-IL17 Mouse Monoclonal Antibody [clone: 6#]

Supplier: CLOUD-CLONE CORP MS
MAA063RB21
MSPP-MA063RB21EA 362 USD
MSPP-MA063RB21
Anti-IL17 Mouse Monoclonal Antibody [clone: 6#]
Antibodies
Monoclonal Antibody to Interleukin 17 (IL17), derived from recombinant IL17 (Val21~Ala153), is reactive with Rabbit.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Interleukin 17 (IL-17; also known as IL-17A) is a 30 to 35 kDa variably glycosylated homodimeric protein that belongs to a unique family of cysteine-knot related proteins. Its sequence was originally isolated from an activated rodent hybridoma and termed CTLA-8. It is synthesized as a 155 amino acid (aa) precursor that contains a 23 aa signal sequence and a 15 kDa, 132 aa mature segment. Although there are two intrachain disulfide bonds that create a ring reminiscent of those found in cysteine-knot proteins, the actual closed knot structure does not appear to form. IL-17 has one potential N-linked glycosylation site. IL-17, a T cell-derived proinf lammatory cytokine, is produced by TCRα/β+CD4-CD8- thymocytes, as well as by activated CD4+ and CD4+CD45RO+ memory T cells.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: IL17
Clonality: Monoclonal
Clone: 6#
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR