Anti-IL12A Mouse Monoclonal Antibody [clone: C3]

Supplier: CLOUD-CLONE CORP MS
MAA059HU24
MSPP-MA059HU24EA 336 USD
MSPP-MA059HU24
Anti-IL12A Mouse Monoclonal Antibody [clone: C3]
Antibodies
Monoclonal Antibody to Interleukin 12A (IL12A), derived from recombinant IL12A (Arg23~Ser219), is reactive with Human/Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Interleukin 12A  also known as IL12A encodes a subunit of a cytokine that acts on T and natural killer cells, and has a broad array of biological activities. The cytokine is a disulfide-linked heterodimer composed of the 35-kD subunit encoded by this gene, and a 40-kD subunit that is a member of the cytokine receptor family. This cytokine is required for the T-cell-independent induction of interferon (IFN)-gamma, and is important for the differentiation of both Th1 and Th2 cells. The responses of lymphocytes to this cytokine are mediated by the activator of transcription protein STAT4. Nitric oxide synthase 2A (NOS2A/NOS2) is found to be required for the signaling process of this cytokine in innate immunity.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: IL12A
Clonality: Monoclonal
Clone: C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR