Anti-IL10 Mouse Monoclonal Antibody [clone: D4]

Supplier: CLOUD-CLONE CORP MS
MAA056RB21
MSPP-MA056RB21EA 362 USD
MSPP-MA056RB21
Anti-IL10 Mouse Monoclonal Antibody [clone: D4]
Antibodies
Monoclonal antibody to Interleukin 10 (IL10), derived from recombinant IL10(Ser19~Ser178), is reactive with Rabbit/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Interleukin-10 is a small protein known as a cytokine that functions as an important regulator of the immune system.  Although IL-10 is known to have many different roles in the immune system, its two major activities include inhibition of cytokine production by macrophages and inhibition of the accessory functions of macrophages during T cell activation. The effects of these actions cause IL-10 to play mainly an anti-inflammatory role in the immune system. IL-10 was originally known as the cytokine synthesis inhibiting factor (CSIF), and the discovery of this protein was based on its biological activity. IL-10 is mainly produced by the Th2 subset of CD4+ helper cells.  However it is also produced by some activated B cells, some Th1 cells, activated macrophages, and some nonhematopoietic sources .

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR