Anti-IGFBP4 Mouse Monoclonal Antibody [clone: C15]

Supplier: CLOUD-CLONE CORP MS
MAA055HU24
MSPP-MA055HU24EA 290 USD
MSPP-MA055HU24
Anti-IGFBP4 Mouse Monoclonal Antibody [clone: C15]
Antibodies
Monoclonal Antibody to Insulin Like Growth Factor Binding Protein 4 (IGFBP4), derived from recombinant IGFBP4 (Asp22~Glu258), is reactive with Human/Mouse/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma in both glycosylated and non-glycosylated forms. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors., IGFBP-4 is a unique protein and it consistently inhibits several cancer cells in vivo and in vitro. Its inhibitory action has been shown in vivo in prostate and colon. It is secreted by all colon cancer cells.The protein itself does not prevent the formation of cancer.However it may reduce the growth of cancer and act as an apoptotic factor.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: IGFBP4
Clonality: Monoclonal
Clone: C15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR