Anti-IFNg Mouse Monoclonal Antibody [clone: 9#]

Supplier: CLOUD-CLONE CORP MS
MAA049RB21
MSPP-MA049RB21EA 362 USD
MSPP-MA049RB21
Anti-IFNg Mouse Monoclonal Antibody [clone: 9#]
Antibodies
Monoclonal antibody to Interferon Gamma (IFNg), derived from recombinant IFNg(Gln24~Tyr167), is reactive with Rabbit/Human/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Interferon gamma (IFN-gama ) is a multifunctional protein first observed as an antiviral activity in cultures of Sindbis virus-infected leukocytes stimulated by PHA . Produced by T lymphocytes and natural killer (NK) cells, IFN-γ is now known to be both an inhibitor of viral replication and a regulator of numerous immunological functions. IFN-γ influences the class of antibody produced by B cells, upregulates class I and II MHC complex antigens and increases. The efficiency of macrophage-mediated killing of intracellular parasites. Most of the activities attributed to IFN-γ are believed to be mediated by IFN-γ-induced proteins. The appearance of such proteins is a consequence of IFN-γ binding to a specific receptor that is distinct from the receptor for IFN-α and β.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: IFNg
Clonality: Monoclonal
Clone: 9#
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Rabbit, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR