Lynx™ PID Temperature Control Set, BriskHeat
Supplier: BRISKHEAT MS
LYNX-SET-JA
MSPP-LYNXSETJAEA 422.31 USD
MSPP-LYNXSETJA
Lynx™ PID Temperature Control Set, BriskHeat
Controllers Temperature Controllers
Lynx™ PID temperature control set with autotuning that controls almost any heater.
  • Everything needed to Plug-and-play
  • 3-button touchpad programming and large 3-digit display
  • Latching alarms for high and low temperature
  • Able to be used with a wide variety of electrical heaters
  • Uses a minimal amount of benchtop space
  • Easy to program and provides precise control
  • Minimum Voltage: 100 V
  • Maximum Voltage: 277 V
It can be used as a single zone controller or be connected together for a system of temperature control.
Maximum continuous ampier load per zone: 7 amp at 25 °C (77 °F) ambient; 4 amp at 40 °C (104 °F)
Ordering information: Each set of temperature controller package includes LYNX temperatue controler module, docking station assembly, 1.8 m power harness, 1.5 m temperature sensor and benchtop stand.