Anti-DBI Rabbit Polyclonal Antibody (Biotinylated)

Supplier: CLOUD-CLONE CORP MS
LAD696MU71
MSPP-LD696MU71EA 396 USD
MSPP-LD696MU71
Anti-DBI Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Biotin-Linked Polyclonal Antibody to Diazepam Binding Inhibitor (DBI), derived from recombinant DBI (Ser2~Ile87), is reactive with Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Acyl-CoA-binding protein isa protein that is regulated by hormones and is involved in lipid metabolism and the displacement of beta-carbolines and benzodiazepines, which modulate signal transduction at type A gamma-aminobutyric acid receptors located in brain synapses. The protein is conserved from yeast to mammals, with the most highly conserved domain consisting of seven contiguous residues that constitute the hydrophobic binding site for medium- and long-chain acyl-Coenzyme A esters. Diazepam binding inhibitor also mediates the feedback regulation of pancreatic secretion and the postprandial release of cholecystokinin, in addition to its role as a mediator in corticotropin-dependent synthesis of steroids in the adrenal gland.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: DBI
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR