Anti-COL7 Mouse Monoclonal Antibody [clone: C9] (Biotinylated)

Supplier: CLOUD-CLONE CORP MS
LAC147HU72
MSPP-LC147HU72EA 407 USD
MSPP-LC147HU72
Anti-COL7 Mouse Monoclonal Antibody [clone: C9] (Biotinylated)
Antibodies
Biotin-Linked Monoclonal Antibody to Collagen Type VII (COL7), derived from recombinant COL7 (Pro190~Asp472), is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Collagen, type VII, alpha 1 encodes the alpha chain of type VII collagen. The type VII collagen fibril, composed of three identical alpha collagen chains, is restricted to the basement zone beneath stratified squamous epithelia. It functions as an anchoring fibril between the external epithelia and the underlying stroma. Mutations in this gene are associated with all forms of dystrophic epidermolysis bullosa. In the absence of mutations, however, an autoimmune response against type VII collagen can result in an acquired form of this disease called epidermolysis bullosa acquisita. Type VII collagen is also found in the retina; its function in this organ is unknown. COL7A1 is located on the short arm of human chromosome 3, in the chromosomal region denoted 3p21.31.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: COL7
Clonality: Monoclonal
Clone: C9
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR