Anti-LGALS3BP Mouse Monoclonal Antibody [clone: D3] (Biotinylated)

Supplier: CLOUD-CLONE CORP MS
LAB766HU72
MSPP-LB766HU72EA 387 USD
MSPP-LB766HU72
Anti-LGALS3BP Mouse Monoclonal Antibody [clone: D3] (Biotinylated)
Antibodies
Biotin-Linked Monoclonal Antibody to Lectin Galactoside Binding, Soluble 3 Binding Protein (LGALS3BP), derived from recombinant LGALS3BP (Met24~Leu221), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Galectin-3-binding protein is a protein encoded by the LGALS3BP gene.The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS3BP has been found elevated in the serum of patients with cancer and in those infected by the human immunodeficiency virus (HIV). It appears to be implicated in immune response associated with natural killer (NK) and lymphokine-activated killer (LAK) cell cytotoxicity. Using fluorescence in situ hybridization the full length 90K cDNA has been localized to chromosome 17q25. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds galectin 1.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: LGALS3BP
Clonality: Monoclonal
Clone: D3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR