LabSand Urine Sample Collection Sand, Braintree Scientific
Supplier: 0000018294
LABSAND-CS
MSPP-LABSANDCSCS 267.07 USD
MSPP-LABSANDCS
LabSand Urine Sample Collection Sand, Braintree Scientific
Animal Supplies Animal Bedding
LabSand is a revolutionary biodegradable hydrophobic sand that has a natural non-toxic urine repelling coating. It allows scientists, researchers, and technicians to quickly and easily collect urine samples without resorting to invasive procedures.
  • Non invasive
  • 100% natural and non-toxic
  • Suitable for testing all common parameters
  • Biodegradable, so disposal is easy
  • Obtain a clean uncontaminated sample
  • Reduces animal stress and maintains high standards of animal welfare
  • Easy collection saves time and makes the process stress free
To use, simply spread the contents of a LabSand bag onto a clean surface and let the animal freely urinate on the sand. The urine will stay on top, allowing you to easily collect the sample with a pipette and vial; or, by dipping a testing strip directly onto the droplet while it is on the sand.
Packaging: Each pack weighs 300 g.