Anti-aHSP Rabbit Polyclonal Antibody (Biotinylated)

Supplier: CLOUD-CLONE CORP MS
LAA412HU71
MSPP-LA412HU71EA 346 USD
MSPP-LA412HU71
Anti-aHSP Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Biotin-Linked Polyclonal Antibody to Alpha-Hemoglobin Stabilizing Protein (aHSP), derived from recombinant aHSP (Met1~Ser102), is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

High pressure induces an irreversible aggregation of the ferrous deoxy α-chains, whereas the AHSP/α-Hb complex shows reversible hexacoordination of the α-Hb without protein aggregation. Upon pressure release, the relaxation kinetics of the transition from the hexacoordinated to pentacoordinated form of α-Hb in the presence of AHSP exhibit a biphasic shape. High pressure did not induce dissociation of α-Hb from its chaperone, as evidenced by the ligand binding kinetics that show a unique rate for the AHSP/α-Hb complex. For both free α-Hb and the AHSP/α-Hb complex, the bimolecular rate constant of CO binding (kCOon) versus pressure exhibits a bell shape, attributed to the transition of the rate-determining step from the chemical barrier to the migration of CO within the protein matrix.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: aHSP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR