General Species LPS ELISA Kit
Supplier: CLOUD-CLONE CORP MS
IEB526GE
MSPP-IEB526GEEA 1067 USD
MSPP-IEB526GE
General Species LPS ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species LPS (Lipopolysaccharide). The assay range is from 0.47 to 30 ng/ml (Sandwich kit) with a sensitivity of 0.19 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
LPS is a major component of the outer membrane of Gram-negative bacteria, contributing greatly to the structural integrity of the bacteria, and protecting the membrane from certain kinds of chemical attack. LPS also increases the negative charge of the cell membrane and helps stabilize the overall membrane structure. It is of crucial importance to gram negative bacteria, whose death results if it is mutated or removed. LPS is an endotoxin, and induces a strong response from normal animal immune systems.LPS acts as the prototypical endotoxin because it binds the CD14/TLR4/MD2 receptor complex, which promotes the secretion of pro-inflammatory cytokines in many cell types, but especially in macrophages. In Immunology, the term 'LPS challenge' refers to the process of exposing a subject to an LPS which may act as a toxin.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.