General Species OT ELISA Kit
Supplier: CLOUD-CLONE CORP MS
HEB052GE
MSPP-HEB052GEEA 710 USD
MSPP-HEB052GE
General Species OT ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species OT (Oxytocin). The assay range is from 1.37 to 1,000 pg/ml (Competitive kit) with a sensitivity of 0.53 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Oxytocin is a mammalian hormone that also acts as a neurotransmitter in the brain.It is best known for its roles in female reproduction: it is released in large amounts after distension of the cervix and vagina during labor, and after stimulation of the nipples, facilitating birth and breastfeeding, respectively. Recent studies have begun to investigate oxytocin's role in various behaviors, including orgasm, social recognition, pair bonding, anxiety, trust, love, and maternal behaviors. Oxytocin is a peptide of nine amino acids (a nonapeptide). The sequence is cysteine - tyrosine - isoleucine - glutamine - asparagine - cysteine - proline - leucine - glycine (CYIQNCPLG). The cysteine residues form a sulfur bridge. Oxytocin has a molecular mass of 1007 daltons. One international unit (IU) of oxytocin is the equivalent of about 2 micrograms of pure peptide.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.