General Species MDA ELISA Kit
Supplier: CLOUD-CLONE CORP MS
HEA597GE
MSPP-HEA597GEEA 876 USD
MSPP-HEA597GE
General Species MDA ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species MDA (Malondialdehyde). The assay range is from 12.35 to 1,000 ng/ml (Competitive kit) with a sensitivity of 4.94 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Malondialdehyde, MDA is a highly reactive three carbon dialdehyde produced as a byproduct of polyunsaturated fatty acid peroxidation and arachidonic acid metabolism. MDA readily combines with several functional groups on molecules including proteins, lipoproteins, and DNA. MDA-modified proteins may show altered physico-chemical behavior and antigenicity. Malondialdehyde is a highly reactive compound that is not typically observed in pure form. In the laboratory it can be generated in situ by hydrolysis of 1,1,3,3-tetramethoxypropane, which is commercially available. It is easily deprotonated to give the sodium salt of the enolate (m.p. 245 °C). Malondialdehyde reacts with deoxyadenosine and deoxyguanosine in DNA, forming DNA adducts, primarily M1G, which is mutagenic. The guanidine group of arginine residues condense with MDA to give 2-aminopyrimidines.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.