Human COL1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
HEA571HU
MSPP-HEA571HUEA 700 USD
MSPP-HEA571HU
Human COL1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human COL1 (Collagen Type I). The assay range is from 15.6 to 1,000 pg/ml (Sandwich kit) with a sensitivity of 5.4 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Collagen is the main protein of connective tissue in animals and the most abundant protein in mammals, making up about 25 to 35% of the whole-body protein content. It is naturally found exclusively in metazoa, including sponges.  In muscle tissue it serves as a major component of endomysium. Collagen constitutes 1 to 2% of muscle tissue, and accounts for 6% of the weight of strong, tendinous muscles. The gelatin used in food and industry is derived from the partial hydrolysis of collagen. is the most abundant collagen of the human body. It is present in scar tissue, the end product when tissue heals by repair. It is found in tendons, skin, artery walls, the endomysium of myofibrils, fibrocartilage, and the organic part of bones and teeth.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.