Human MMP1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
HEA097HU
MSPP-HEA097HUEA 700 USD
MSPP-HEA097HU
Human MMP1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human MMP1 (Matrix Metalloproteinase 1). The assay range is from 15.6 to 1,000 pg/ml (Sandwich kit) with a sensitivity of 5.8 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
MMP-1 is produced by fibroblasts, chondrocytes, macrophages, endothelial cells, and osteoblasts. It is induced by the pro-inflammatory cytokines IL-1 and TNF-α, various growth factors such as EGF, PDGF, FGF basic, and Oncostatin M, chemical agents such as cAMP and phorbol esters and events occurring at the cell surface such as cell fusion and phagocytosis. MMP-1 plays a significant role in the degradation of different types of collagen in extracellular matrix remodeling. It is implicated in a variety of processes involving collagen degradation such as emphysema, atherosclerosis, rheumatoid and osteoarthritis, periodontal and respiratory disease, angiogenesis and tumorigenesis, tissue remodeling and wound healing, and inflammatory bowel disease.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.